3-Chlor-2,6-dibrom-4-methylanilin
Lieferant: FLUOROCHEM
Synonyme:
3-Chlor-2,6-dibrom-p-toluidin
018364-5G
018364-25G
FLUO018364-5GEA
43.1
EUR
FLUO018364-5G
FLUO018364-25G
3-Chlor-2,6-dibrom-4-methylanilin
3-Chlor-2,6-dibrom-4-methylanilin
Formel:
C₇H₆Br₂ClN Molecular Weight: 299.39 |
MDL:
MFCD00051580 CAS-Nummer: 84483-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...