4,4,5,5,6,6,7,7,7-Nonafluor-1-heptanol
Lieferant: FLUOROCHEM
019068-25G
019068-5G
FLUO019068-25GEA
0
EUR
FLUO019068-25G
FLUO019068-5G
4,4,5,5,6,6,7,7,7-Nonafluor-1-heptanol
4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol
Formel:
C₇H₇F₉O |
MDL:
MFCD02183552 CAS-Nummer: 83310-97-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...