2-Brom-4-fluor-6-methylanilin

Lieferant: FLUOROCHEM

Synonyme: 6-Brom-4-fluor-o-toluidin

023891-25G 023891-5G
FLUO023891-25GEA 152 EUR
FLUO023891-25G FLUO023891-5G
2-Brom-4-fluor-6-methylanilin
2-Brom-4-fluor-6-methylanilin
Formel: C₇H₇BrFN
Molecular Weight: 204.04 g/mol
MDL: MFCD01861196
CAS-Nummer: 202865-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR