1,4-Pentadien-3-ol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Hydroxy-1,4-pentadien, Penta-1,4-dien-3-ol
324663-25G
324663-5G
SIAL324663-25GEA
362
EUR
SIAL324663-25G
SIAL324663-5G
1,4-Pentadien-3-ol, Sigma-Aldrich®
1,4-Pentadien-3-ol
Formel:
C₅H₈O Molecular Weight: 84,12 g/mol |
MDL:
MFCD00008633 CAS-Nummer: 922-65-6 EINECS: 213-080-9 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...