1-Butylimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Butyl-1H-imidazol, N-Butylimidazol, 1-Butyl-1H-imidazol

M-Clarity™ Qualitätsstufe = MQ100

348414-100G 348414-25G
SIAL348414-100GEA 104 EUR
SIAL348414-100G SIAL348414-25G
1-Butylimidazol, Sigma-Aldrich®
1-Butylimidazole
Formel: C₇H₁₂N₂
Molecular Weight: 124,19 g/mol
MDL: MFCD00042753
CAS-Nummer: 4316-42-1
EINECS: 224-335-9
UN: 2690
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR