3-(Trimethylsilyl)propiolsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Trimethylsilyl)acetylencarbonsäure, 3-(Trimethylsilyl)propinsäure

M-Clarity™ Qualitätsstufe = MQ100

403482-1G 403482-5G
SIAL403482-1GEA 58.2 EUR
SIAL403482-1G SIAL403482-5G
3-(Trimethylsilyl)propiolsäure, Sigma-Aldrich®
3-(Trimethylsilyl)propiolsäure
Formel: C₆H₁₀O₂Si
Molecular Weight: 142,23 g/mol
MDL: MFCD00190216
CAS-Nummer: 5683-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR