1H,1H,2H,2H-Heptadecafluor-1-decanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluor-1-decanol, 1H,1H,2H,2H-Perfluor-1-decanol

M-Clarity™ Qualitätsstufe = MQ200

532789-25G 532789-5G
SIAL532789-25GEA 153 EUR
SIAL532789-25G SIAL532789-5G
1H,1H,2H,2H-Heptadecafluor-1-decanol, Sigma-Aldrich®
1H,1H,2H,2H-Heptadecafluor-1-decanol
Formel: C₁₀H₅F₁₇O
Molecular Weight: 464,12 g/mol
MDL: MFCD00039544
CAS-Nummer: 678-39-7
EINECS: 211-648-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR