2-Biphenylyl(dicyclohexyl)phosphin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-Biphenyl)dicyclohexylphosphin

M-Clarity™ Qualitätsstufe = MQ100

638099-5G 638099-1G 638099-25G
SIAL638099-5GEA 92.7 EUR
SIAL638099-5G SIAL638099-1G SIAL638099-25G
2-Biphenylyl(dicyclohexyl)phosphin, Sigma-Aldrich®
2-(Dicyclohexylphosphino)biphenyl
Formel: C₂₄H₃₁P
Molecular Weight: 350,48 g/mol
MDL: MFCD01862441
CAS-Nummer: 247940-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR