3'-Chlor-4'-methoxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(3-Chlor-4-methoxyphenyl)ethan-1-on, 1-(3-Chlor-4-methoxyphenyl)ethanon

M-Clarity™ Qualitätsstufe = MQ200

648809-1G 648809-5G
SIAL648809-1GEA 39.6 EUR
SIAL648809-1G SIAL648809-5G
3'-Chlor-4'-methoxyacetophenon, Sigma-Aldrich®
3'-Chlor-4'-methoxyacetophenon
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
MDL: MFCD04973413
CAS-Nummer: 37612-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR