2-Brom-4-fluor-6-methylanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Brom-4-fluor-o-toluidin

M-Clarity™ Qualitätsstufe = MQ100

669369-10G 669369-1G
SIAL669369-10GEA 108 EUR
SIAL669369-10G SIAL669369-1G
2-Brom-4-fluor-6-methylanilin, Sigma-Aldrich®
2-Brom-4-fluor-6-methylanilin
Formel: C₇H₇BrFN
Molecular Weight: 204.04 g/mol
MDL: MFCD01861196
CAS-Nummer: 202865-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR