Kalium(brommethyl)trifluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (Brommethyl)trifluorborsäure Kaliumsalz

M-Clarity™ Qualitätsstufe = MQ100

681342-1G 681342-5G
SIAL681342-1GEA 66.6 EUR
SIAL681342-1G SIAL681342-5G
Kalium(brommethyl)trifluoroborat, Sigma-Aldrich®
Kalium(brommethyl)trifluoroborat
Formel: CH₂BBrF₃K
Molecular Weight: 200,83 g/mol
MDL: MFCD09265154
CAS-Nummer: 888711-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR