1-(Triisopropylsilyl)-1-propin

Lieferant: CHEMPUR SILANE

Synonyme: (1-Propinyl)triisopropylsilan

SILS18024.1 SILS18024.5
SILNSILS18024.1EA 0 EUR
SILNSILS18024.1 SILNSILS18024.5
1-(Triisopropylsilyl)-1-propin
1-(Triisopropylsilyl)-1-propin
Formel: C₁₂H₂₄Si
Molecular Weight: 196,41 g/mol
MDL: MFCD00015636
CAS-Nummer: 82192-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR