1-(Trimethylsilyl)-1-propin
Lieferant: CHEMPUR SILANE
SILS20225.5
SILS20225.25
SILNSILS20225.5EA
0
EUR
SILNSILS20225.5
SILNSILS20225.25
1-(Trimethylsilyl)-1-propin
1-(Trimethylsilyl)-1-propin
Formel:
C₆H₁₂Si Siedepunkt: 99 °C (1013 hPa) Dichte: 0,759 g/cm³ (20 °C) |
MDL:
MFCD00009271 CAS-Nummer: 6224-91-5 EINECS: 228-314-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...