Propargyltrimethylsilan

Lieferant: CHEMPUR SILANE

Synonyme: (2-Propinyl)trimethylsilan, 3-(Trimethylsilyl)propin

SILS20227.5 SILS20227.25
SILNSILS20227.5EA 0 EUR
SILNSILS20227.5 SILNSILS20227.25
Propargyltrimethylsilan
Propargyltrimethylsilan
Formel: C₆H₁₂Si
Molecular Weight: 112,25 g/mol
MDL: MFCD00042922
CAS-Nummer: 13361-64-3
EINECS: 236-427-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR