2-Biphenylyl(dicyclohexyl)phosphin, weiße Kristalle

Lieferant: ABCR

Synonyme: (2-Biphenyl)dicyclohexylphosphin

S15-1140.0002 AB130655.0010
ABCRS15-1140.0002EA 0 EUR
ABCRS15-1140.0002 ABCRAB130655.0010
2-Biphenylyl(dicyclohexyl)phosphin, weiße Kristalle
2-(Dicyclohexylphosphino)biphenyl
Formel: C₂₄H₃₁P
Molecular Weight: 350,48 g/mol
MDL: MFCD01862441
CAS-Nummer: 247940-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR