2-Brom-2-methylbutan

Lieferant: FLUOROCHEM

005914-10G 005914-25G
FLUO005914-10GEA 0 EUR
FLUO005914-10G FLUO005914-25G
2-Brom-2-methylbutan
2-Brom-2-methylbutan
Formel: C₅H₁₁Br
Molecular Weight: 151,05 g/mol
Siedepunkt: 108 °C (1013 hPa)
Dichte: 1,2439 g/cm³ (0 °C)
MDL: MFCD00000127
CAS-Nummer: 507-36-8
EINECS: 208-071-1
UN: 1993
ADR: 3,II
Merck Index: 12,00644

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR