2,6-Dimethyl-3,5-heptandion ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

42005.03 42005.06
42005.03EA 63.8 EUR
42005.03 42005.06
2,6-Dimethyl-3,5-heptandion ≥98%
2,6-Dimethyl-3,5-heptandion

Purity: 98+% CAS No.: 18362-64-6 Molecular Formula: C9H16O2 Formula Weight: 156.22 Physical Form: Liquid Density: 0.91 UN No.: 1224 MDL No.: MFCD00015040


Metal chelating agent; preparation of metal diketonates for MOCVD

Formel: C₉H₁₆O₂
Siedepunkt: 212 °C (1013 hPa)
Dichte: 0,91
Lagertemperatur: Raumtemperatur
MDL: MFCD00015040
CAS-Nummer: 18362-64-6
EINECS: 242-234-8
UN: 1224
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR