5,5-Dimethylacetylaceton ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5,5-Dimethylhexane-2,4-dione, 2,2-Dimethyl-3,5-hexanedione, 5,5-Diméthylhexanedione-2,4

L13104.03 L13104.06
L13104.03EA 69.2 EUR
L13104.03 L13104.06
5,5-Dimethylacetylaceton ≥99%
5,5-Dimethylacetylaceton

Purity: 99% CAS No.: 7307-04-2 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 57°(134°F) Refractive Index: 1.4600 Boiling Point: 60°/10mm Beilstein Reference: 969813 UN No.: 1224 MDL No.: MFCD00142767

Formel: C₈H₁₄O₂
Molecular Weight: 142,2 g/mol
Siedepunkt: 60 °C (10 mmHg)
Flammpunkt: 57 °C (134 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00142767
CAS-Nummer: 7307-04-2
EINECS: 230-760-0
UN: 1224
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR