1-Chlor-3-methylbutan ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-Methylbutylchloride
L11371.09
L11371.18
L11371.09EA
32.2
EUR
L11371.09
L11371.18
1-Chlor-3-methylbutan ≥98%
1-Chlor-3-methylbutan
Purity: 98%
CAS No.: 107-84-6
Molecular Formula: C5H11Cl
Formula Weight: 106.60
Flash Point: 16°(60°F)
Refractive Index: 1.4090
Melting Point: -104°
Boiling Point: 99-100°
Beilstein Reference: 1730988
Merck Reference: 14,5116
UN No.: 1107
MDL No.: MFCD00013693
Formel:
C₅H₁₁Cl Molecular Weight: 106,6 g/mol Siedepunkt: 99…100 °C Schmelzpunkt: –104 °C Dichte: 0,880 Flammpunkt: 16 °C (60 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013693 CAS-Nummer: 107-84-6 EINECS: 203-525-5 UN: 1107 ADR: 3,II Merck Index: 14,5116 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...