1,1,1-Trifluor-5,5-dimethyl-2,4-hexandion
Lieferant: Thermo Scientific
Synonyme:
5,5-Dimethyl-1,1,1-trifluorohexane-2,4-dione
RJC04016.1
RJC04016.250
MAYBRJC04016.1EA
0
EUR
MAYBRJC04016.1
CHMPRJC04016.250
1,1,1-Trifluor-5,5-dimethyl-2,4-hexandion
1,1,1-Trifluor-5,5-dimethyl-2,4-hexandion
Formel:
C₈H₁₁F₃O₂ Molecular Weight: 196,17 g/mol Siedepunkt: 178 °C (1013 hPa) Dichte: 1,129 g/cm³ (20 °C) |
MDL:
MFCD00040841 CAS-Nummer: 22767-90-4 EINECS: 245-211-0 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...