Isobutylamin 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Methylpropylamin, 1-Amino-2-methylpropan
A11017.AK
A11017.AU
A11017.AKEA
26.2
EUR
A11017.AK
A11017.AU
Isobutylamin 99%
Isobutylamin
Purity: 99%
CAS No.: 78-81-9
Molecular Formula: (CH3)2CHCH2NH2
Formula Weight: 73.14
Flash Point: -9°(15°F)
Refractive Index: 1.3970
Odour: Amine-like
Melting Point: -85°
Boiling Point: 67-69°
Beilstein Reference: 385626
Merck Reference: 14,5132
UN No.: 1214
MDL No.: MFCD00008146
Formel:
C₄H₁₁N Molecular Weight: 73,14 g/mol Siedepunkt: 67…69 °C Schmelzpunkt: –85 °C Dichte: 0,736 Flammpunkt: –9 °C (15 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008146 CAS-Nummer: 78-81-9 EINECS: 201-145-4 UN: 1214 ADR: 3,II Merck Index: 14,5132 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...