2,2,4-Trimethylpentan 99+% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Isooktan, Isobutyltrimethylmethan
31787.AP
31787.K2
31787.M1
31787.APEA
41.8
EUR
31787.AP
31787.K2
31787.M1
2,2,4-Trimethylpentan 99+% ACS
Isooctan
Purity: 99+%
CAS No.: 540-84-1
Molecular Formula: (CH3)2CHCH2C(CH3)3
Formula Weight: 114.23
Flash Point: -7°(19°F)
Physical Form: Liquid
Density: 0.692
Refractive Index: 1.3910
Odour: Nearly Odourless
Melting Point: -107°
Boiling Point: 98-99°
Solubility: Soluble in benzene, toluene, xylene, chloroform, ether, CS2, CCl4, and DMF
Beilstein Reference: 1696876
Merck Reference: 14,5193
UN No.: 1262
MDL No.: MFCD00008943
Solvent, thinner, in determination of fuel octane numbers
Formel:
(CH₃)₂CHCH₂C(CH₃)₃ Siedepunkt: 98…99 °C Schmelzpunkt: –107 °C Dichte: 0,692 Flammpunkt: –7 °C (19 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008943 CAS-Nummer: 540-84-1 EINECS: 208-759-1 UN: 1262 ADR: 3,II Merck Index: 14,5193 |