2,3-Dimethylbutan ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
B22223.06
B22223.14
B22223.22
B22223.06EA
32.9
EUR
B22223.06
B22223.14
B22223.22
2,3-Dimethylbutan ≥99%
2,3-Dimethylbutane
Purity: 99%
CAS No.: 79-29-8
Molecular Formula: C6H14
Formula Weight: 86.18
Flash Point: -29°(-20°F)
Refractive Index: 1.375
Odour: Mild
Melting Point: -129°
Boiling Point: 57-59°
Beilstein Reference: 1730737
UN No.: 2457
MDL No.: MFCD00008925
Formel:
C₆H₁₄ Molecular Weight: 86,18 g/mol Siedepunkt: 57…59 °C Schmelzpunkt: –129 °C Dichte: 0,662 Flammpunkt: –29 °C (–20 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008925 CAS-Nummer: 79-29-8 EINECS: 201-193-6 UN: 2457 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...