6-Brom-1-hexen 95%

Lieferant: Thermo Scientific
Achtung

L12909.03 L12909.06
L12909.03EA 38.4 EUR
L12909.03 L12909.06
6-Brom-1-hexen 95%
6-Brom-1-hexen

Purity: 95% CAS No.: 2695-47-8 Molecular Formula: C6H11Br Formula Weight: 163.06 Flash Point: 54°(129°F) Refractive Index: 1.4652 Odour: Characteristic Boiling Point: 147° Beilstein Reference: 506203 UN No.: 1993 MDL No.: MFCD00000269

Formel: C₆H₁₁Br
Molecular Weight: 163,06 g/mol
Siedepunkt: 147 °C
Dichte: 1,217
Flammpunkt: 54 °C (129 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000269
CAS-Nummer: 2695-47-8
EINECS: 220-267-9
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR