4-Brom-3,3,4,4-tetrafluor-1-buten
Lieferant: FLUOROCHEM
004278-1G
004278-5G
FLUO004278-1GEA
34.1
EUR
FLUO004278-1G
FLUO004278-5G
4-Brom-3,3,4,4-tetrafluor-1-buten
4-Brom-3,3,4,4-tetrafluor-1-buten
Formel:
C₄H₃BrF₄ Molecular Weight: 206.97 g/mol Siedepunkt: 57 Dichte: 1.627 g/cm³ (20 °C) |
MDL:
MFCD00042189 CAS-Nummer: 18599-22-9 EINECS: 242-440-8 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...