3,4-Epoxy-1-buten ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Butadiene monoxide

L00238.03 L00238.06 L00238.14
L00238.03EA 55.2 EUR
L00238.03 L00238.06 L00238.14
3,4-Epoxy-1-buten ≥98%
3,4-Epoxy-1-buten

Purity: 98% CAS No.: 930-22-3 Molecular Formula: C4H6O Formula Weight: 70.09 Storage Temperature: 2°C to 8°C Flash Point: -50°(-58°F) Refractive Index: 1.4180 Odour: Recognizable Melting Point: -135° Boiling Point: 65-66° Beilstein Reference: 103170 UN No.: 1993 MDL No.: MFCD00005149

Formel: C₄H₆O
Molecular Weight: 70,09 g/mol
Siedepunkt: 65…66 °C
Schmelzpunkt: –135 °C
Dichte: 0,870
Flammpunkt: –50 °C (–58 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00005149
CAS-Nummer: 930-22-3
EINECS: 213-210-4
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR