cis-4-Decenal 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(Z)-4-Decenal
L06382.03
L06382.06
L06382.03EA
28
EUR
L06382.03
L06382.06
cis-4-Decenal 95%
cis-4-Decenal
Purity: 95%
CAS No.: 21662-09-9
Molecular Formula: C10H18O
Formula Weight: 154.25
Storage Temperature: 2°C to 8°C
Flash Point: 81°(177°F)
Refractive Index: 1.4440
Boiling Point: 78-80°/10mm
Beilstein Reference: 2323646
MDL No.: MFCD00007024
Formel:
C₁₀H₁₈O Molecular Weight: 154,25 g/mol Siedepunkt: 78…80 °C (10 mmHg) Dichte: 0,857 Flammpunkt: 81 °C (177 °F) |
MDL:
MFCD00007024 CAS-Nummer: 21662-09-9 EINECS: 244-514-5 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...