trans-2-Decenal 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(E)-2-Decenal
L05901.06
L05901.14
L05901.06EA
55.7
EUR
L05901.06
L05901.14
trans-2-Decenal 95%
trans-2-Decenal
Purity: 95%
CAS No.: 3913-81-3
Molecular Formula: C10H18O
Formula Weight: 154.25
Storage Temperature: 2°C to 8°C
Flash Point: 93°(199°F)
Refractive Index: 1.4565
Boiling Point: 117°/17mm
Beilstein Reference: 1704445
MDL No.: MFCD00014679
Formel:
C₁₀H₁₈O Molecular Weight: 154,25 g/mol Siedepunkt: 117 °C (17 mmHg) Dichte: 0,840 Flammpunkt: 93 °C (199 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00014679 CAS-Nummer: 3913-81-3 EINECS: 223-474-2 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...