trans-2-Pentenal 96%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(E)-2-Pentenal
L06108.06
L06108.14
L06108.06EA
38.5
EUR
L06108.06
L06108.14
trans-2-Pentenal 96%
trans-2-Pentenal
Purity: 96%
CAS No.: 1576-87-0
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 22°(71°F)
Refractive Index: 1.4430
Odour: Fruit-like
Boiling Point: 125°
Beilstein Reference: 1719742
UN No.: 1989
MDL No.: MFCD00009615
Formel:
C₅H₈O Molecular Weight: 84,12 g/mol Siedepunkt: 125 °C Dichte: 0,860 Flammpunkt: 22 °C (71 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009615 CAS-Nummer: 1576-87-0 EINECS: 216-414-1 UN: 1989 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...