3-Methoxypropen ≥94%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Allyl methyl ether

18652.09 18652.18
18652.09EA 97.8 EUR
18652.09 18652.18
3-Methoxypropen ≥94%
3-Methoxypropen

Purity: 94% CAS No.: 627-40-7 Molecular Formula: CH2=CHCH2OCH3 Formula Weight: 72.10 Flash Point: -23°(-94°F) Physical Form: Liquid Density: 0.77 Refractive Index: 1.3803 Odour: Ether-like Boiling Point: 42-43° UN No.: 3271 MDL No.: MFCD00008649

Formel: C₄H₈O
Molecular Weight: 72,11 g/mol
Siedepunkt: 42…43 °C
Dichte: 0,77
Flammpunkt: –23 °C (–94 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008649
CAS-Nummer: 627-40-7
EINECS: 210-997-6
UN: 3271
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR