Acroleindiethylacetal 96%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Acroleinacetal, 3,3-Diethoxypropen
B24821.09
B24821.18
B24821.09EA
31.2
EUR
B24821.09
B24821.18
Acroleindiethylacetal 96%
Acroleindiethylacetal
Purity: 96%
CAS No.: 3054-95-3
Molecular Formula: CH2=CHCH(OCH2CH3)2
Formula Weight: 130.19
Flash Point: 13°(55°F)
Refractive Index: 1.4010
Boiling Point: 124-125°
Beilstein Reference: 1701567
UN No.: 2374
MDL No.: MFCD00009239
Formel:
C₇H₁₄O₂ Molecular Weight: 130,19 g/mol Siedepunkt: 124…125 °C Schmelzpunkt: –77 °C Dichte: 0,852 Flammpunkt: 13 °C (55 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009239 CAS-Nummer: 3054-95-3 EINECS: 221-276-0 UN: 2374 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...