Tiglinaldehyd ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: trans-2,3-Dimethylacrolein, (E)-2-Methyl-2-butenal

L08113.06 L08113.14
L08113.06EA 42.6 EUR
L08113.06 L08113.14
Tiglinaldehyd ≥97%
trans-2-Methyl-2-butenal

Purity: 97% CAS No.: 497-03-0 Molecular Formula: C5H8O Formula Weight: 84.12 Storage Temperature: 2°C to 8°C Flash Point: 18°(64°F) Refractive Index: 1.4480 Odour: Fruit-like Boiling Point: 117-119° Beilstein Reference: 1698207 UN No.: 1989 MDL No.: MFCD00006977

Formel: C₅H₈O
Molecular Weight: 84,12 g/mol
Siedepunkt: 117…119 °C
Dichte: 0,871
Flammpunkt: 18 °C (64 °F)
MDL: MFCD00006977
CAS-Nummer: 497-03-0
EINECS: 207-833-0
UN: 1989
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR