9-Vinylanthracen ≥97%

Lieferant: Thermo Scientific

B22333.03 B22333.06
B22333.03EA 62.4 EUR
B22333.03 B22333.06
9-Vinylanthracen ≥97%
9-Vinylanthracen

Purity: 97% CAS No.: 2444-68-0 Molecular Formula: C16H12 Formula Weight: 204.27 Storage Temperature: 2°C to 8°C Melting Point: 62-65° Boiling Point: 61-66°/10mm Beilstein Reference: 2041641 MDL No.: MFCD00003576

Formel: C₁₆H₁₂
Molecular Weight: 204,27 g/mol
Siedepunkt: 61…66 °C (10 mmHg)
Schmelzpunkt: 62…65 °C
Dichte: 1,269 g/cm³ (–150.15 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00003576
CAS-Nummer: 2444-68-0
EINECS: 219-486-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR