1,3-Dibrombenzol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
A12723.14
A12723.22
A12723.36
A12723.14EA
54.6
EUR
A12723.14
A12723.22
A12723.36
1,3-Dibrombenzol ≥97%
1,3-Dibrombenzol
Purity: 97+%
CAS No.: 108-36-1
Molecular Formula: C6H4Br2
Formula Weight: 235.91
Flash Point: 93°(199°F)
Refractive Index: 1.6080
Odour: Odourless
Melting Point: -7°
Boiling Point: 219-220°
Beilstein Reference: 1904538
UN No.: 3082
MDL No.: MFCD00000078
Formel:
C₆H₄Br₂ Molecular Weight: 235,91 g/mol Siedepunkt: 219…220 °C Schmelzpunkt: –7 °C Dichte: 1,955 Flammpunkt: 93 °C (199 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000078 CAS-Nummer: 108-36-1 EINECS: 203-574-2 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...