3-Brombenzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

A11941.22 A11941.14 A11941.36
A11941.22EA 82.4 EUR
A11941.22 A11941.14 A11941.36
3-Brombenzaldehyd ≥97%
3-Brombenzaldehyd

Purity: 97% CAS No.: 3132-99-8 Molecular Formula: C7H5BrO Formula Weight: 185.03 Flash Point: 96°(204°F) Refractive Index: 1.5940 Odour: Characteristic Melting Point: 18-21° Boiling Point: 234-236° Beilstein Reference: 1634534 MDL No.: MFCD00003345

Formel: C₇H₅BrO
Molecular Weight: 185,02 g/mol
Siedepunkt: 234…236 °C
Schmelzpunkt: 18…21 °C
Dichte: 1,580
Flammpunkt: 96 °C (204 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003345
CAS-Nummer: 3132-99-8
EINECS: 221-526-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR