5-Brom-o-anisaldehyd ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Bromo-2-methoxybenzaldehyde, 5-Bromo-2-anisaldehyde

A19396.14 A19396.22 AB125716.0100
A19396.14EA 19.1 EUR
A19396.14 A19396.22 ABCRAB125716.0100
5-Brom-o-anisaldehyd ≥98%
5-Brom-o-anisaldehyd

Purity: 98+% CAS No.: 25016-01-7 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 114-118° Beilstein Reference: 508840

Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
Siedepunkt: 269 °C (1013 hPa)
Schmelzpunkt: 114…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003347
CAS-Nummer: 25016-01-7
EINECS: 246-564-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR