5-Brom-o-anisaldehyd
Lieferant: MENAI
Synonyme:
5-Bromo-2-methoxybenzaldehyde, 5-Bromo-2-anisaldehyde
IP13.50
IP13.100
MENAIP13.50EA
0
EUR
MENAIP13.50
MENAIP13.100
5-Brom-o-anisaldehyd
5-Brom-o-anisaldehyd
Formel:
C₈H₇BrO₂ Molecular Weight: 215,05 g/mol Siedepunkt: 269 °C (1013 hPa) Schmelzpunkt: 117 °C |
MDL:
MFCD00003347 CAS-Nummer: 25016-01-7 EINECS: 246-564-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...