6-Bromveratraldehyd 97

Lieferant: Thermo Fisher Scientific

Synonyme: 6-Bromo-3,4-dimethoxybenzaldehyde, 2-Bromo-4,5-dimethoxybenzaldehyde

A13591.06 A13591.14 A13591.22
A13591.06EA 57.5 EUR
A13591.06 A13591.14 A13591.22
6-Bromveratraldehyd 97
6-Bromveratraldehyd

Purity: 97% CAS No.: 5392-10-9 Molecular Formula: C9H9BrO3 Formula Weight: 245.08 Melting Point: 147-152° Beilstein Reference: 2052355

Formel: C₉H₉BrO₃
Molecular Weight: 245.07 g/mol
Siedepunkt: 300
Schmelzpunkt: 147…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003301
CAS-Nummer: 5392-10-9
EINECS: 226-390-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR