3,5-Dibromsalicylaldehyd
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Carbaldehyd-4,6-dibromphenol
BTB02697.1
BTB02697EA
BTB02697EE
MAYBBTB02697.1EA
0
EUR
MAYBBTB02697.1
MAYBBTB02697EA
MAYBBTB02697EE
3,5-Dibromsalicylaldehyd
3,5-Dibromsalicylaldehyd
Formel:
C₇H₄Br₂O₂ Molecular Weight: 279,92 g/mol Siedepunkt: 261 °C (1013 hPa) Schmelzpunkt: 86 °C |
MDL:
MFCD00003318 CAS-Nummer: 90-59-5 EINECS: 202-003-4 Merck Index: 13,03052 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...