3,5-Dibrom-4-hydroxybenzaldehyd
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
4-Carbaldehyd-2,6-dibromphenol
RH00089.250
RH00089.1
CHMPRH00089.250EA
0
EUR
CHMPRH00089.250
MAYBRH00089.1
3,5-Dibrom-4-hydroxybenzaldehyd
3,5-Dibrom-4-hydroxybenzaldehyd
Formel:
C₇H₄Br₂O₂ Molecular Weight: 279,92 g/mol |
MDL:
MFCD00016980 CAS-Nummer: 2973-77-5 EINECS: 221-017-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...