2-Brombenzoesäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: o-Bromobenzoic Acid

A13608.0B A13608.18 A13608.30
A13608.0BEA 523 EUR
A13608.0B A13608.18 A13608.30
2-Brombenzoesäure ≥98%
2-Brombenzoesäure

Purity: 98% CAS No.: 88-65-3 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Melting Point: 146-150° Beilstein Reference: 971266

Formel: C₇H₅BrO₂
Molecular Weight: 201,02 g/mol
Siedepunkt: 296 °C (1013 hPa)
Schmelzpunkt: 146…150 °C
Dichte: 1,930
Flammpunkt: >100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002402
CAS-Nummer: 88-65-3
EINECS: 201-848-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR