3,5-Dibromanthranilsäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-3,5-dibromobenzoicacid, 2-Amino-3,5-dibromobenzoic acid

L07419.06 L07419.14
L07419.06EA 39.3 EUR
L07419.06 L07419.14
3,5-Dibromanthranilsäure 98%
3,5-Dibromanthranilsäure

Purity: 98% CAS No.: 609-85-8 Molecular Formula: C7H5Br2NO2 Formula Weight: 294.93 Melting Point: 230-235° Beilstein Reference: 779291

Formel: C₇H₅Br₂NO₂
Molecular Weight: 294,93 g/mol
Schmelzpunkt: 230…235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017087
CAS-Nummer: 609-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR