2-Brom-5-methoxybenzoesäure
Lieferant: Thermo Scientific
SB00527.1
SB00527.10
SB00527.50
MAYBSB00527.1EA
0
EUR
MAYBSB00527.1
MAYBSB00527.10
MAYBSB00527.50
2-Brom-5-methoxybenzoesäure
2-Bromo-5-methoxybenzoic acid
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol |
MDL:
MFCD00020214 CAS-Nummer: 22921-68-2 EINECS: 245-329-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...