3,5-Dichlor-4-hydroxybenzoesäure ≥97%

Lieferant: Thermo Scientific
Achtung

A12923.14 A12923.06 A12923.22
A12923.14EA 90.4 EUR
A12923.14 A12923.06 A12923.22
3,5-Dichlor-4-hydroxybenzoesäure ≥97%
3,5-Dichloro-4-hydroxybenzoic acid

Purity: 97% CAS No.: 3336-41-2 Molecular Formula: C7H4Cl2O3 Formula Weight: 207.01 Melting Point: 265-270° Beilstein Reference: 2616297

Formel: C₇H₄Cl₂O₃
Molecular Weight: 207,01 g/mol
Siedepunkt: 341 °C (1013 hPa)
Schmelzpunkt: 265…270 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002550
CAS-Nummer: 3336-41-2
EINECS: 222-071-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR