3,5-Dibrom-4-hydroxybenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A18361.14 A18361.22
A18361.14EA 59.5 EUR
A18361.14 A18361.22
3,5-Dibrom-4-hydroxybenzoesäure ≥98%
3,5-Dibromo-4-hydroxybenzoic acid

Purity: 98% CAS No.: 3337-62-0 Molecular Formula: C7H4Br2O3 Formula Weight: 295.93 Melting Point: 269-273° Beilstein Reference: 2416147

Formel: C₇H₄Br₂O₃
Molecular Weight: 295.91 g/mol
Siedepunkt: 365
Schmelzpunkt: 269…273 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002548
CAS-Nummer: 3337-62-0
EINECS: 222-075-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR