3-Brom-4-hydroxybenzoesäure ≥98%

Lieferant: Thermo Scientific

B21338.09 B21338.18
B21338.09EA 42.3 EUR
B21338.09 B21338.18
3-Brom-4-hydroxybenzoesäure ≥98%
3-Bromo-4-hydroxybenzoic acid

Purity: 98% CAS No.: 14348-41-5 Molecular Formula: C7H5BrO3 Formula Weight: 217.02 Melting Point: 176-178° MDL No.: MFCD00017547

Formel: C₇H₅BrO₃
Molecular Weight: 217,02 g/mol
Schmelzpunkt: 176…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017547
CAS-Nummer: 14348-41-5
EINECS: 238-299-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR