3-Hydroxy-2,4,6-tribrombenzoesäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,4,6-Tribromo-3-hydroxybenzoic acid

334890050 334890250
ACRO334890050EA 81.4 EUR
ACRO334890050 ACRO334890250
3-Hydroxy-2,4,6-tribrombenzoesäure 97%
3-Hydroxy-2,4,6-tribromobenzoic acid
Formel: C₇H₃Br₃O₃
Molecular Weight: 374.81 g/mol
Siedepunkt: 396
Schmelzpunkt: 144…149 °C
MDL: MFCD00055557
CAS-Nummer: 14348-40-4
UN: 2811

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 144 to 149 °C
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR