3-Hydroxy-2,4,6-tribrombenzoesäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2,4,6-Tribromo-3-hydroxybenzoic acid
B23790.06
B23790.14
B23790.06EA
70.3
EUR
B23790.06
B23790.14
3-Hydroxy-2,4,6-tribrombenzoesäure ≥98%
3-Hydroxy-2,4,6-tribromobenzoic acid
Purity: 98%
CAS No.: 14348-40-4
Molecular Formula: C7H3Br3O3
Formula Weight: 374.81
Melting Point: 144-149°
Beilstein Reference: 2844497
Formel:
C₇H₃Br₃O₃ Molecular Weight: 374.81 g/mol Siedepunkt: 396 Schmelzpunkt: 144…149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055557 CAS-Nummer: 14348-40-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...