1-Brom-2,4-dichlorbenzol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2,4-Dichlorobromobenzene
018266-5G
018266-10G
FLUO018266-5GEA
0
EUR
FLUO018266-5G
FLUO018266-10G
1-Brom-2,4-dichlorbenzol
1-Brom-2,4-dichlorbenzol
Formel:
C₆H₃BrCl₂ Molecular Weight: 225,9 g/mol |
MDL:
MFCD00018333 CAS-Nummer: 1193-72-2 EINECS: 214-778-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...