4-Brom-1,2-dichlorbenzol 98+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Bromo-3,4-dichlorobenzene
269810250
269811000
ACRO269810250EA
56.2
EUR
ACRO269810250
ACRO269811000
4-Brom-1,2-dichlorbenzol 98+%
4-Brom-1,2-dichlorbenzol
Formel:
C₆H₃BrCl₂ Molecular Weight: 225,9 g/mol Siedepunkt: 140…142 °C (30 mmHg) Schmelzpunkt: 23…24 °C Dichte: 1,781 g/cm³ (20 °C) |
MDL:
MFCD00040849 CAS-Nummer: 18282-59-2 EINECS: 242-160-6 UN: 2810 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.5986 to 1.6006 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...